Learn More
BIOHIPPO Mouse CAPN10 Microsample
SDP
Catalog No. 5000315127
Encompass
Mouse CAPN10 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CAPN10 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 106 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CAPN10 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CAPN10 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CAPN10 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15127 Supplier BIOHIPPO Supplier No. BHE15210937
Mouse CAPN10 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CAPN10 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 106 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CAPN10 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CAPN10 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CAPN10 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color