Learn More
Sino Biological Mouse Carbonic Anhydrase IV/CA4 HEK293 Overexpression Lysate
Catalog No. NC3761041
Encompass_Preferred
This Mouse Carbonic Anhydrase IV/CA4 overexpression lysate was created in HEK293 Cells and intented for use as a Western blot (WB) positive control. Purification of Carbonic Anhydrase IV/CA4 protein (Cat: 50350-M08H) from the overexpression lysate was verified.
Catalog No. NC3761041 Supplier Sino Biological Supplier No. 50350M08HL
This Mouse Carbonic Anhydrase IV/CA4 overexpression lysate was created in HEK293 Cells and intented for use as a Western blot (WB) positive control. Purification of Carbonic Anhydrase IV/CA4 protein (Cat: 50350-M08H) from the overexpression lysate was verified.