Learn More
Biomatik Corporation Mouse Carbonic Anhydrase II (CA2) ELISA Kit
Supplier:  Biomatik Corporation EKU10704
Encompass_Preferred
Mouse Carbonic Anhydrase II (CA2) ELISA Kit for detecting CA2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00920. Gene ID: 12349. Target Synonyms: CA-II; CAII; Car2; CAC; Carbonate dehydratase II; Carbonic anhydrase C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7114