Learn More
ACROBiosystems ACROBiosystems Mouse Carbonic Anhydrase IX / CA9 (32-390) Protein, His Tag
Catalog No. 502106475
Encompass_Preferred
Recombinant Protein;50UG;Mouse Carbonic Anhydrase IX / CA9 (32-390), His Tag (CA9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Asp 390 (Accession # Q8VHB5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.7 kDa.
Catalog No. 50-210-6475 Supplier ACROBiosystems Supplier No. CA9M52H350UG
Recombinant Protein;50UG;Mouse Carbonic Anhydrase IX / CA9 (32-390), His Tag (CA9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Asp 390 (Accession # Q8VHB5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.7 kDa.