Learn More
Biomatik Corporation Mouse Carbonic Anhydrase IX (CA9) ELISA Kit
Supplier:  Biomatik Corporation EKU02923
Encompass_Preferred
Mouse Carbonic Anhydrase IX (CA9) ELISA Kit for detecting CA9 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.65pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VHB5. Gene ID: 230099. Target Synonyms: CA-IX; CAIX; MN; pMW1; P54/58N; Carbonic Dehydratase; Renal cell carcinoma-associated antigen G250; RCC-Associated Protein G250; Carbonate dehydratase IX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7196