Learn More
Biomatik Corporation Mouse Carboxypeptidase A3 (CPA3) ELISA Kit
Supplier:  Biomatik Corporation EKU02934
Encompass_Preferred
Mouse Carboxypeptidase A3 (CPA3) ELISA Kit for detecting CPA3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15089. Gene ID: 12873. Target Synonyms: MC-CPA; Mast cell carboxypeptidase A; Carboxypeptidase A3, Mast Cell. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6405