Learn More
Biomatik Corporation Mouse Carboxypeptidase B1, Tissue (CPB1) ELISA Kit
Supplier:  Biomatik Corporation EKU08304
Encompass_Preferred
Mouse Carboxypeptidase B1, Tissue (CPB1) ELISA Kit for detecting CPB1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 2.52pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2RS76. Gene ID: 76703. Target Synonyms: CPB; PCPB; PASP; Pancreas-specific protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6439