Learn More
American Research Products Inc Mouse CarboXyterminal propepti
Supplier:  American Research Products Inc CSBE08080M
Encompass
Mouse CarboXyterminal propeptide of type procollagen P CP ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26262