Learn More
American Research Products Inc Mouse cardiac troponin I cTn
Supplier:  American Research Products Inc CSBE08421MIS
Encompass
Mouse cardiac troponin I (cTn-I) ELISA kit from CUSABIO Detection Range 15 6pg/mL-1000pg/mL Sensitivity 7 8pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26129