Learn More
Biomatik Corporation Mouse Cardiotrophin 1 (CT1) ELISA Kit
Supplier:  Biomatik Corporation EKU02953
Encompass_Preferred
Mouse Cardiotrophin 1 (CT1) ELISA Kit for detecting CT1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60753. Gene ID: 13019. Target Synonyms: CTF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7200