Learn More
Biomatik Corporation Mouse Carnitine Acetyltransferase (CRAT) ELISA Kit
Supplier:  Biomatik Corporation EKU02958
Encompass_Preferred
Mouse Carnitine Acetyltransferase (CRAT) ELISA Kit for detecting CRAT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47934. Gene ID: 12908. Target Synonyms: CAT1; Carnitine O-Acetyltransferase; Carnitine acetylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9073