Learn More
World Precision Instrument Mouse Carotid Artery Catheter, 82 mm, 32 G
SDP
Catalog No. 502539398
Encompass
Catheters used for sampling or dosing can vary and depend on access or animal size. We carry catheters used for common applications. All catheters are ETO sterilized.
Do you need a custom design? Tubing diameters, material, beads, shapes and lengths of existing products can be modified as necessary. Call us with your requirements.
Catalog No. 50-253-9398 Supplier World Precision Instrument Supplier No. 504143
Catheters used for sampling or dosing can vary and depend on access or animal size. We carry catheters used for common applications. All catheters are ETO sterilized.
Do you need a custom design? Tubing diameters, material, beads, shapes and lengths of existing products can be modified as necessary. Call us with your requirements.