Learn More
American Research Products Inc Mouse Cartilage oligomeric mat
Catalog No. 5000327931
Encompass
Mouse Cartilage oligomeric matriX protein (COMP) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27931 Supplier American Research Products Inc Supplier No. CSBEL005778MO
Mouse Cartilage oligomeric matriX protein (COMP) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates Method Sandwich