Learn More
Biomatik Corporation Mouse Caspase 14 (CASP14) ELISA Kit
Supplier:  Biomatik Corporation EKU10085
Encompass_Preferred
Mouse Caspase 14 (CASP14) ELISA Kit for detecting CASP14 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O89094. Gene ID: 12365. Target Synonyms: Caspase 14 Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7218