Learn More
Cusabio Technology LLC Mouse Caspase 3Casp-3 ELISA
Catalog No. 5000049311
Encompass_Preferred
This kit detects CASP3 in mouse serum plasma and tissue homogenates using a sandwich ELISA method offering quantitative measurement within a dynamic range of 0 312 ng/mL to 20 ng/mL With a sensitivity of 0 078 ng/mL it enables precise detection of low-abundance targets making it suitable for apoptosis-related research The assay requires a short incubation period of 1-5 hours and utilizes sample volumes between 50-100 L ensuring efficient workflow compatibility Absorbance is measured at 450 nm for result interpretation Designed for reliability and reproducibility this kit provides researchers with a robust tool for analyzing caspase-3 activity in diverse biological samples
Catalog No. 50-000-49311 Supplier Cusabio Technology LLC Supplier No. CSBE08858M
This kit detects CASP3 in mouse serum plasma and tissue homogenates using a sandwich ELISA method offering quantitative measurement within a dynamic range of 0 312 ng/mL to 20 ng/mL With a sensitivity of 0 078 ng/mL it enables precise detection of low-abundance targets making it suitable for apoptosis-related research The assay requires a short incubation period of 1-5 hours and utilizes sample volumes between 50-100 L ensuring efficient workflow compatibility Absorbance is measured at 450 nm for result interpretation Designed for reliability and reproducibility this kit provides researchers with a robust tool for analyzing caspase-3 activity in diverse biological samples