Learn More
Sino Biological Recombinant Mouse 5'-nucleotidase/CD73/NT5E Protein (His Tag)
Supplier:  Sino Biological 50231M08H50
Encompass_Preferred
A DNA sequence encoding the mouse NT5E (NP_035981.1) without the propeptide, corresponding to the amino acid (Met 1-Lys 549) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-224-6788