Learn More
ACROBiosystems Mouse Cathepsin L 20ug
Catalog No. 502535414
Encompass_Preferred
Mouse Cathepsin L Protein His Tag (CAL-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 18 - Asn 334 (Accession # P06797-1).
Catalog No. 50-253-5414 Supplier ACROBiosystems Supplier No. CALM52H320UG
Mouse Cathepsin L Protein His Tag (CAL-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 18 - Asn 334 (Accession # P06797-1).