Learn More
Biomatik Corporation Mouse Cathepsin L (CTSL) ELISA Kit
Supplier:  Biomatik Corporation EKU03038
Encompass_Preferred
Mouse Cathepsin L (CTSL) ELISA Kit for detecting CTSL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06797. Gene ID: 13039. Target Synonyms: CTSL1; CATL; CTS-L; MEP; Cathepsin L1; Major excreted protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4571