Learn More
Sino Biological Recombinant Mouse CD39/ENTPD1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50398M08B50
Encompass_Preferred
A DNA sequence encoding the mouse ENTPD1 (P55772) extracellular domain (Thr 38-Ile 478) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-224-6966