Learn More
American Research Products Inc Mouse Cathepsin L1 CTSL1 ELI
Catalog No. 5000328131
Encompass
Mouse Cathepsin L1 (CTSL1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28131 Supplier American Research Products Inc Supplier No. CSBEL006193MO
Mouse Cathepsin L1 (CTSL1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich