Learn More
Alzet ALZETTM Mouse Intraperitoneal Catheter: Optimized for Intraperitoneal Delivery from any ALZET Osmotic Pump
SDP
Catalog No. NC0484203
Encompass
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous delivery of test agents to the mouse peritoneal space. EtO sterilized and ready for immediate use.Catheter Material: Silicone and polyurethaneSpecifications: 6 cm total length; distal tip 3 Fr; internal volume ~15 μlOptimum Animal Size: Adult mouseFeatures & Benefits:•Fenestrations over silicone tip for enhanced distribution•Movable suture bead for optimum adjustment of inserted catheter segment•Flexible silicone segment minimizes trauma•Stress relief catheter junction prevents catheter kinking and improves patency•Length adjustable by trimming ALZET connection segment•ALZET connection provides secure fit to any ALZET pump
Catalog No. NC0484203 Supplier Alzet Supplier No. 0007771
This ALZET catheter connects to any ALZET Osmotic Pump to enable continuous delivery of test agents to the mouse peritoneal space. EtO sterilized and ready for immediate use.Catheter Material: Silicone and polyurethaneSpecifications: 6 cm total length; distal tip 3 Fr; internal volume ~15 μlOptimum Animal Size: Adult mouseFeatures & Benefits:•Fenestrations over silicone tip for enhanced distribution•Movable suture bead for optimum adjustment of inserted catheter segment•Flexible silicone segment minimizes trauma•Stress relief catheter junction prevents catheter kinking and improves patency•Length adjustable by trimming ALZET connection segment•ALZET connection provides secure fit to any ALZET pump