Learn More
SAI Infusion Technologies Mouse Jugular Vein Catheter - 10cm total length
SDP
Catalog No. NC0880995
Encompass
Jugular Vein Catheter for Mice. Polyurethane, 1cm 2Fr tip, 3F body, 2 suture beads, 10cm total length. Access with 22G blunt needle (such as B22-50). Sterile. Catheters may be used for sampling or infusion. Our biocompatible polyurethane catheters are designed to enhance patency and minimize vessel trauma during implantation. Retention (suture) beads are accurately positioned from the catheter tip for optimal implantation position within the vessel. Beads are also positioned to provide a secure tension loop to maximize continued patency during movement and growth. Product is for laboratory animal research only.
Catalog No. NC0880995 Supplier SAI Infusion Technologies Supplier No. MJVC3F
Jugular Vein Catheter for Mice. Polyurethane, 1cm 2Fr tip, 3F body, 2 suture beads, 10cm total length. Access with 22G blunt needle (such as B22-50). Sterile. Catheters may be used for sampling or infusion. Our biocompatible polyurethane catheters are designed to enhance patency and minimize vessel trauma during implantation. Retention (suture) beads are accurately positioned from the catheter tip for optimal implantation position within the vessel. Beads are also positioned to provide a secure tension loop to maximize continued patency during movement and growth. Product is for laboratory animal research only.