Learn More
Sino Biological Anti-Caveolin-3/CAV3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106397T44100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse Caveolin-3/CAV3, and purified by antigen affinity chromatography.
Catalog No. 50-241-1217