Learn More
BIOHIPPO Mouse CC16 Microsample
SDP
Catalog No. 5000315500
Encompass
Mouse CC16 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CC16 in serum plasma lung lavage fluid tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 02 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CC16 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CC16 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CC16 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15500 Supplier BIOHIPPO Supplier No. BHE15212649
Mouse CC16 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CC16 in serum plasma lung lavage fluid tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 02 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CC16 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CC16 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CC16 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color