Learn More
Kingfisher Biotech Inc Rabbit LIF ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1230M004
Encompass_Preferred
The Rabbit Leukemia Inhibitory Factor (LIF) ELISA contains capture antibody, standard, and detection antibody for development of a Rabbit Leukemia Inhibitory Factor ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include LIF, CDF, DIA, HILDA, MLPLI, leukemia inhibitory factor, interleukin 6 family cytokine, LIF interleukin 6 family cytokine. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5378