Learn More
Proteintech Group Inc Mouse CCL11/Eotaxin ELISA
SDP
Supplier:  Proteintech Group Inc KE1004396T
Encompass_Preferred
CCL11 also named Eotaxin, was initially discovered as an eosinophil-specific chemoattractant. CCL11 plays a central role in both allergic and non-allergic inflammatory reactions by recruiting immune cells such as eosinophils,basophils and Th2 lymphocytes. CCL11 binds to the chemokine receptors CCR2, CCR3 and CCR5, with highest affinity to CCR3. High levels of CCL11 have been described in several chronic inflammatory diseases, such as allergic rhinitis, atopic dermatitis, asthma, gastrointestinal disease and rheumatoid arthritis.
Catalog No. 50-197-4167