Learn More
Sino Biological Anti-TARC/CCL17 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50046RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CCL17 / TARC (rM CCL17 / TARC; Catalog#50046-MNAB; AAD56601.1; Met1-Pro93). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1770