Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit

Catalog No. p-7145531
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SMJE00B 1 Pack (6 Plates)
MJE00B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SMJE00B Supplier R&D Systems™ Supplier No. SMJE00B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse CCL2/JE/MCP-1 Quantikine ELISA Kit quantifies mouse CCL2/JE/MCP-1 in cell culture supernates (50 ul), tissue lysates (50 ul), tissue culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

1 Kit

Specifications

Assay Range 7.81 - 500 pg/mL
Assay Sensitivity 0.666 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Lysates
Specificity Natural and Recombinant Mouse MCP-1
For Use With (Application) ELISA
Gene ID (Entrez) 6347
Gene Symbol CCL2
Host Species Mouse
Quantity 1 Pack (6 Plates)
Sample Volume Cell Culture Supernates (50 uL),Tissue Lysates (50 uL),Tissue Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target CCL2/JE/MCP-1
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.