Learn More
Sino Biological Recombinant Anti-MCP-1/CCL2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50368R01720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse MCP-1/CCL2 (rM MCP-1/CCL2; Catalog#50368-M08B; NP_035463.1; Met1-Asn148).
Catalog No. 50-240-9032