Learn More
Bon Opus Biosciences Recombinant Mouse CCL20 (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP104250UG
Encompass_Preferred
Recombinant Mouse CCL20 (C-mFc) is produced by Human Cells. The target gene encoding Ala28-Met97 is expressed with a C-mFc tag. Accession# NP_058656.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1042
Catalog No. 50-217-2641