Learn More
Sino Biological Anti-MDC/CCL22 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51135RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CCL22 / MDC (rM CCL22 / MDC; Catalog#51135-M07E; O88430; Gly26-Ser92). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8688