Learn More
Sino Biological Recombinant Mouse MDC/CCL22 Protein (His Tag)
Supplier:  Sino Biological 51135M07E1
Encompass_Preferred
A DNA sequence encoding the mouse CCL22 (O88430) (Gly26-Ser92) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-2984