Learn More
Sino Biological Anti-MIP-1 alpha/CCL3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51114RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CCL3 / Mip1a (Catalog#51114-MNAE; P10855; Ala24-Ala92). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8260