Learn More
BIOHIPPO Mouse CCL3L1 Microsample
SDP
Catalog No. 5000314234
Encompass
Mouse CCL3L1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CCL3L1 in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 37 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CCL3L1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CCL3L1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CCL3L1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14234 Supplier BIOHIPPO Supplier No. BHE15212241
Mouse CCL3L1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CCL3L1 in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 37 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CCL3L1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CCL3L1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CCL3L1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color