Learn More
Proteintech Group Inc Mouse CCL4 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE1003096T
Encompass_Preferred
CCL4(C-C chemokine ligand 4), also known as MIP-1β, plays a key role in inflammation and immune regulation. CCL4 is a major HIV-suppressive factor produced by CD8+ T cells. CCL4 is produced by a wide variety of cells including: immune cells (monocytes, B and T cells), fibroblasts, endothelial, and epithelial cells. CCL4 has a broad spectrum of target cells including immature dendritic cells, which express the cognate receptor CCR5. Increased expression of CCL4 was reported in the monocytes and blood of elderly individuals. Concentration of CCL4 within the body increases with age, which may cause chronic inflammation and liver damage.
Catalog No. 50-197-4154