Learn More
Raybiotech Inc Mouse CCL6 ELISA Kit
Supplier:  Raybiotech Inc ELMCCL61
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 18 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Detect mouse CCL6, no cross reactivity with recombinant mouse CCL3, CCL4, or CCL9
Catalog No. 50-194-9939