Learn More
Sino Biological Recombinant Mouse MCP-3/CCL7 Protein (His Tag)
Supplier:  Sino Biological 50627M07E1
Encompass_Preferred
A DNA sequence encoding the mouse Ccl7 (NP_038682.1) (Gln24-Pro97) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3186