Learn More
Sino Biological Recombinant Mouse MCP-2/CCL8 Protein (His & NusA Tag)
Supplier:  Sino Biological 50181M24E1
Encompass_Preferred
A DNA sequence encoding the mature form of mouse CCL8 (Q9Z121) (Gly 24-Pro 94) was fused with the polyhistidine-tagged NusA tag at the N-terminus.
Catalog No. 50-255-3387