Learn More
Lonza Walkersville Mouse CD-1 Hepatocytes Cryo
Catalog No. 5000496305
Encompass
Cryopreserved CD-1 mouse hepatocytes suspension format specifically pre-tested for metabolic activity of general Phase I and Phase II metabolism in suspension This product is not suitable for applications requiring adherent cells
Catalog No. 50-004-96305 Supplier Lonza Walkersville Supplier No. MCCS01
Cryopreserved CD-1 mouse hepatocytes suspension format specifically pre-tested for metabolic activity of general Phase I and Phase II metabolism in suspension This product is not suitable for applications requiring adherent cells