Learn More
Lonza Walkersville Mouse CD-1 Hepatocytes, Cryopreserved, Plateable
Catalog No. NC1844928
Encompass
All our plateable cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension. They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture.Applications:Drug metabolism in plated monolayer formatSpecies-species comparisonsGenomicsContent:Cryo vial, > 5 million viable cells
Catalog No. NC1844928 Supplier Lonza Walkersville Supplier No. MCCP01
All our plateable cryopreserved primary animal hepatocytes are specifically pre tested for metabolic activity of general Phase I and Phase II metabolism in suspension. They are tested for healthy morphology lasting 72 hours in monlayer sandwich culture.Applications:Drug metabolism in plated monolayer formatSpecies-species comparisonsGenomicsContent:Cryo vial, > 5 million viable cells