Learn More
Discovery Life Sciences Corning Gentest™ Mouse (CD-1) Pooled Liver Microsomes, Male, 0.5 mL
SDP
Catalog No. NC1801066
Encompass
Tissue fractions from human and animal livers represent an important tool in preclinical metabolism studies for predicting the toxicity and pharmacokinetic properties of a drug compound. Liver microsomes from humans and animal species play an important role in evaluating drug compounds for metabolic stability, reactive metabolites, drug-drug interactions, reaction phenotyping, and metabolite identification.
Catalog No. NC1801066 Supplier Discovery Life Sciences Supplier No. 452701
Tissue fractions from human and animal livers represent an important tool in preclinical metabolism studies for predicting the toxicity and pharmacokinetic properties of a drug compound. Liver microsomes from humans and animal species play an important role in evaluating drug compounds for metabolic stability, reactive metabolites, drug-drug interactions, reaction phenotyping, and metabolite identification.