Learn More
Lonza Walkersville Mouse CD1 Brain Cortex Neurons
Catalog No. 5000496366
Encompass
Ready-to-use Clonetics Mouse Brain CD1 Cortex Neurons (M-CX-400) are isolated from Mouse CD1 brain (E14 15) These are sold as cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods and are ready for immediate culture Each vial of cortex cells contains approximately 4 million viable cells This will seed into 10 or more wells of a 24-well plate and 50 or more wells of a 96-well plate using the recommended plating densities and medium After approximately 4 days in culture the cells will form a neurite network and by the 7th day debris will be minimal Recommended medium PNGM BulletKit (CC-4461)
Catalog No. 50-004-96366 Supplier Lonza Walkersville Supplier No. MCX400
Ready-to-use Clonetics Mouse Brain CD1 Cortex Neurons (M-CX-400) are isolated from Mouse CD1 brain (E14 15) These are sold as cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods and are ready for immediate culture Each vial of cortex cells contains approximately 4 million viable cells This will seed into 10 or more wells of a 24-well plate and 50 or more wells of a 96-well plate using the recommended plating densities and medium After approximately 4 days in culture the cells will form a neurite network and by the 7th day debris will be minimal Recommended medium PNGM BulletKit (CC-4461)