Learn More
Lonza Walkersville Mouse CD1 Brain Mixed Astrocyt
Catalog No. 5000496385
Encompass
Ready-to-use Clonetics Mouse Brain Mixed Astrocytes are obtained from CD1 mouse brain passaged once and prepared as cell suspensions for shipment Each vial contains approximately 1 million cells The astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated The cells are shipped overnight on dry ice Recommended medium AGM BulletKit (CC-3186)
Catalog No. 50-004-96385 Supplier Lonza Walkersville Supplier No. MASM430
Ready-to-use Clonetics Mouse Brain Mixed Astrocytes are obtained from CD1 mouse brain passaged once and prepared as cell suspensions for shipment Each vial contains approximately 1 million cells The astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated The cells are shipped overnight on dry ice Recommended medium AGM BulletKit (CC-3186)