Learn More
Lonza Walkersville Mouse CD1 Brain Striatum Neuro
Catalog No. 5000496419
Encompass
Ready-to-use Clonetics Mouse Brain Striatum Neurons are isolated from Mouse CD1 brain (E14 15) The cell suspensions of high quality primary embryonic brain neuronal cells (including glia) are prepared by standardized methods and are ready for immediate culture Each vial of striatum cells contains approximately 4 million viable cells This will seed into 6 or more wells of a 24-well plate and 30 or more wells of a 96-well plate using the recommended plating densities and medium After approximately 4 days in culture the cells will form a neurite network and by the 7th day debris will be minimal Recommended Media PNGM BulletKit (CC-4461)
Catalog No. 50-004-96419 Supplier Lonza Walkersville Supplier No. MCP402
Ready-to-use Clonetics Mouse Brain Striatum Neurons are isolated from Mouse CD1 brain (E14 15) The cell suspensions of high quality primary embryonic brain neuronal cells (including glia) are prepared by standardized methods and are ready for immediate culture Each vial of striatum cells contains approximately 4 million viable cells This will seed into 6 or more wells of a 24-well plate and 30 or more wells of a 96-well plate using the recommended plating densities and medium After approximately 4 days in culture the cells will form a neurite network and by the 7th day debris will be minimal Recommended Media PNGM BulletKit (CC-4461)