Learn More
Zyagen Labs Mouse CD1 Lymph nodes Total Protein, 0.5mg
SDP
Catalog No. NC3585209
Encompass_Preferred
Mouse Lymph nodes total protein (protein lysate) is isolated from freshly harvested tissues of single healthy normal donor. Tissues are homogenized in protein lysis buffer supplemented with a cocktail of 6 mammalian protease inhibitors to minimize proteolysis, precisely quantified, and stored at -80oC.
Catalog No. NC3585209 Supplier Zyagen Labs Supplier No. MT703
Mouse Lymph nodes total protein (protein lysate) is isolated from freshly harvested tissues of single healthy normal donor. Tissues are homogenized in protein lysis buffer supplemented with a cocktail of 6 mammalian protease inhibitors to minimize proteolysis, precisely quantified, and stored at -80oC.