Learn More
Zyagen Labs Mouse CD1 Lymph nodes Paraffin Sections, 10 slides
SDP
Catalog No. NC3585090
Encompass_Preferred
Mouse Lymph Nodes paraffin sections are made of freshly harvested tissues, fixed in 10% neutral buffered formalin, and processed for paraffin embedding. Paraffin?blocks are sectioned at a thickness of 5microns and mounted on positively charged slides (2-4 sections per slide depending on tissue size).
Catalog No. NC3585090 Supplier Zyagen Labs Supplier No. MP703
Mouse Lymph Nodes paraffin sections are made of freshly harvested tissues, fixed in 10% neutral buffered formalin, and processed for paraffin embedding. Paraffin?blocks are sectioned at a thickness of 5microns and mounted on positively charged slides (2-4 sections per slide depending on tissue size).