Learn More
Innovative Research Inc Mouse CD1 Red Blood Cells 25ml
Catalog No. 502034898
Encompass_Preferred
Mouse CD1 Red Blood Cells from Innovative Research are manufactured using whole blood collections from type CD1 mice. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4898 Supplier Innovative Research Inc Supplier No. IMSCD1RBC25ML
Mouse CD1 Red Blood Cells from Innovative Research are manufactured using whole blood collections from type CD1 mice. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.