Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) 3CLpro / 3C-like protease-His & AVI Recombinant Protein
Supplier:  Sino Biological 40594V56E100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) 3CL Protease (YP_009725295.1) (Ser3264-Gln3569) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.
Catalog No. 50-224-7241