Learn More
Sino Biological Recombinant Anti-CD14 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50023R21720
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD14 (rM CD14; Catalog#50023-M08H; NP_033971.1; Met1-Pro345).
Catalog No. 50-241-1386