Learn More
Sino Biological Recombinant Anti-CD14 Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 50023R503C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD14 (rM CD14; Catalog#50023-M08H; NP_033971.1; Met1-Pro345) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-8317